Thermo Scientific Chemicals

1-Propanol, for HPLC, Thermo Scientific Chemicals

Catalog number: 389600010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

1-Propanol, for HPLC, Thermo Scientific Chemicals

Catalog number: 389600010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 389600010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
71-23-8
IUPAC Name
propan-1-ol
Molecular Formula
C3H8O
InChI Key
BDERNNFJNOPAEC-UHFFFAOYSA-N
SMILES
CCCO
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Acidity
=<0.0002 meq/g
Water
=<0.05 %
GC
>=99.9 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text