2-(Dicyclohexylphosphino)-2'-isopropylbiphenyl, 98%, Thermo Scientific Chemicals
2-(Dicyclohexylphosphino)-2'-isopropylbiphenyl, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(Dicyclohexylphosphino)-2'-isopropylbiphenyl, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Packaging:
Glass bottle
Catalog number 389720010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS251320-85-1
IUPAC Namedicyclohexyl[2'-(propan-2-yl)-[1,1'-biphenyl]-2-yl]phosphane
Molecular FormulaC27H37P
InChI KeyPQWWLQRSCJJOIS-UHFFFAOYSA-N
SMILESCC(C)C1=CC=CC=C1C1=CC=CC=C1P(C1CCCCC1)C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=97.5 %
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only