1x1 image pixel for data collection
Allyl methyl carbonate, 98%
Allyl methyl carbonate, 98%
Allyl methyl carbonate, 98%
Allyl methyl carbonate, 98%
Thermo Scientific Chemicals

Allyl methyl carbonate, 98%

CAS: 35466-83-2 | C5H8O3 | 116.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 389750250
Price (USD)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS35466-83-2
IUPAC Namemethyl prop-2-en-1-yl carbonate
Molecular FormulaC5H8O3
InChI KeyYHLVIDQQTOMBGN-UHFFFAOYSA-N
SMILESCOC(=O)OCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4050 to 1.4090 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Clear colorless to almost colorless
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link