Calcium carbide, 72-82%
Calcium carbide, 72-82%
Calcium carbide, 72-82%
Calcium carbide, 72-82%
Thermo Scientific Chemicals

Calcium carbide, 72-82%

CAS: 75-20-7 | C2Ca | 64.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
250 g
1 kg
Packaging:
Glass bottle
Catalog number 389790025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS75-20-7
IUPAC Namecalcium acetylene
Molecular FormulaC2Ca
InChI KeyUIXRSLJINYRGFQ-UHFFFAOYSA-N
SMILES[Ca++].[C-]#[C-]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric12 to 23 % (CaO)
Particle size7 to 25 mm
Appearance (Color)Dark grey
Appearance (Form)Lumps
Titration Complexometric72 to 82 % (CaC2)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link