Thermo Scientific Chemicals

Zinc sulfate monohydrate, 99%, Thermo Scientific Chemicals

Catalog number: 389800010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Zinc sulfate monohydrate, 99%, Thermo Scientific Chemicals

Catalog number: 389800010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 389800010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7446-19-7
IUPAC Name
zinc(2+) hydrate sulfate
Molecular Formula
H2O5SZn
InChI Key
RNZCSKGULNFAMC-UHFFFAOYSA-L
SMILES
O.[Zn++].[O-]S([O-])(=O)=O
Appearance (Color)
White
Appearance (Form)
Powder
Titration Complexometric
98.5 to 101.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text