Thermo Scientific Chemicals

Sodium phosphate, tribasic, pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 389810025
2.5 kg, Plastic Drum, Each
Thermo Scientific Chemicals

Sodium phosphate, tribasic, pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 389810025
2.5 kg, Plastic Drum, Each
Quantity
Packaging
Catalog number: 389810025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7601-54-9
IUPAC Name
trisodium phosphate
Molecular Formula
Na3O4P
InChI Key
RYFMWSXOAZQYPI-UHFFFAOYSA-K
SMILES
[Na+].[Na+].[Na+].[O-]P([O-])([O-])=O
Appearance (Form)
Crystalline powder
Titration with HCl
>=96.0 %
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text