Thermo Scientific Chemicals

Chromium(III) chloride tetrahydrofuran complex (1:3), 98%, Thermo Scientific Chemicals

Catalog number: 389850010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Chromium(III) chloride tetrahydrofuran complex (1:3), 98%, Thermo Scientific Chemicals

Catalog number: 389850010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 389850010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10170-68-0
IUPAC Name
tris(oxolane); trichlorochromium
Molecular Formula
C12H24Cl3CrO3
InChI Key
CYOMBOLDXZUMBU-UHFFFAOYSA-K
SMILES
Cl[Cr](Cl)Cl.C1CCOC1.C1CCOC1.C1CCOC1
Appearance (Color)
Purple
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with Na2S2O3
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text