Thermo Scientific Chemicals

Sodium phosphate, monobasic, 99%, for analysis, anhydrous, Thermo Scientific Chemicals

Catalog number: 389870100
10 kg, Plastic drum, Each
Thermo Scientific Chemicals

Sodium phosphate, monobasic, 99%, for analysis, anhydrous, Thermo Scientific Chemicals

Catalog number: 389870100
10 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 389870100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7558-80-7
IUPAC Name
sodium dihydrogen phosphate
Molecular Formula
H2NaO4P
InChI Key
AJPJDKMHJJGVTQ-UHFFFAOYSA-M
SMILES
[Na+].OP(O)([O-])=O
Appearance (Color)
White
Appearance (Form)
Crystalline powder
Titration with NaOH
>=98.5 %
pH
4.1 to 4.7 (5 % solution)
Insoluble matter
=<0.05 % (in water)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text