Palladium(II) nitrate hydrate, Thermo Scientific Chemicals
Palladium(II) nitrate hydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Palladium(II) nitrate hydrate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 389910010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namepalladium(2+) dinitrate
Molecular FormulaN2O6Pd
InChI KeyGPNDARIEYHPYAY-UHFFFAOYSA-N
SMILES[Pd++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Molecular Weight (g/mol)230.43
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Appearance (Color)Brown to brown-black
Palladium (Pd)39 to 46 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only