Mercury(I) sulfate, 97%, Thermo Scientific Chemicals
Mercury(I) sulfate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Mercury(I) sulfate, 97%, Thermo Scientific Chemicals

Have Questions?
Quantity:
100 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 389921000
Price (USD)/ Each
221.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
261.00 
Save 39.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
221.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
261.00 
Save 39.35 (15%)
Add to cart
Mercury(I) sulfate, 97%, Thermo Scientific Chemicals
Catalog number389921000
Price (USD)/ Each
221.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
261.00 
Save 39.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7783-36-0
IUPAC Namemercury(2+) sulfate
Molecular FormulaHgO4S
InChI KeyDOBUSJIVSSJEDA-UHFFFAOYSA-L
SMILES[Hg++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to beige
Titration Iodimetric96.0 to 104.0 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only