Thermo Scientific Chemicals

Sodium borohydride, 12% solution in 40% aq. sodium hydroxide solution, Thermo Scientific Chemicals

Catalog number: 389930025
2.5 L, Plastic bottle, Each
Thermo Scientific Chemicals

Sodium borohydride, 12% solution in 40% aq. sodium hydroxide solution, Thermo Scientific Chemicals

Catalog number: 389930025
2.5 L, Plastic bottle, Each
Quantity
Packaging
Catalog number: 389930025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16940-66-2, 1310-73-2, 67-56-1, 7732-18-5
Appearance (Color)
Colorless
Appearance (Form)
Liquid
Titration Iodometric
11 to 13 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text