Amberlite™ FPC3500, ion-exchange resin, Thermo Scientific Chemicals
Amberlite™ FPC3500, ion-exchange resin, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Amberlite™ FPC3500, ion-exchange resin, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 389992500
Price (USD)/ Each
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
MDL NumberMFCD08460528
CAS12125-02-9, 7732-18-5
SpecificationsSpecification SheetSpecification Sheet
Harmonic mean size0.450 to 0.650 mm
Moisture holding capacity60 to 70 % (H)
Appearance (Form)Beads
Appearance (Color)White to light yellow
Uniformity coefficient=<2.00
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resin
RUO – Research Use Only