Chemical Identifiers
CAS1332-09-8
IUPAC Name2-[9b-fluoro-10-hydroxy-2,9a,11a-trimethyl-7-oxo-1-(propanoyloxy)-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl propanoate
Molecular FormulaC28H37FO7
InChI KeyCIWBQSYVNNPZIQ-UHFFFAOYNA-N
SMILESCCC(=O)OCC(=O)C1(OC(=O)CC)C(C)CC2C3CCC4=CC(=O)C=CC4(C)C3(F)C(O)CC12C
View more