Ammonium hydroxide, for analysis, 20% solution in water
Ammonium hydroxide, for analysis, 20% solution in water
Ammonium hydroxide, for analysis, 20% solution in water
Ammonium hydroxide, for analysis, 20% solution in water
Thermo Scientific Chemicals

Ammonium hydroxide, for analysis, 20% solution in water

CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
View all
Glass Bottle
Catalog number 390070025
Price (USD)
107.00
Each
In stock
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
107.00
Each
Add to cart
Chemical Identifiers
CAS1336-21-6
IUPAC Nameamine hydrate
Molecular FormulaH5NO
InChI KeyVHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILESN.O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Aluminium (Al)=<0.00001 %
Calcium (Ca)=<0.0001 %
Chromium (Cr)=<0.000005 %
Gallium (Ga)=<0.000005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link