Thermo Scientific Chemicals

Calcium citrate tribasic tetrahydrate, 98+%, for analysis, Thermo Scientific Chemicals

Catalog number: 390080010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Calcium citrate tribasic tetrahydrate, 98+%, for analysis, Thermo Scientific Chemicals

Catalog number: 390080010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 390080010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5785-44-4
IUPAC Name
tricalcium bis(2-hydroxypropane-1,2,3-tricarboxylate) tetrahydrate
Molecular Formula
C12H18Ca3O18
InChI Key
LNIZKKFWMDARJV-UHFFFAOYSA-H
SMILES
O.O.O.O.[Ca++].[Ca++].[Ca++].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Appearance (Color)
White
Appearance (Form)
Powder
Infrared spectrum
Conforms
Titration Complexometric
98.0 to 102.0 % (on dried basis)
Loss on drying
10.0 % to 13.3 % (150°C, 4 h)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text