Calcium citrate tribasic tetrahydrate, 98+%, for analysis
Calcium citrate tribasic tetrahydrate, 98+%, for analysis
Calcium citrate tribasic tetrahydrate, 98+%, for analysis
Calcium citrate tribasic tetrahydrate, 98+%, for analysis
Thermo Scientific Chemicals

Calcium citrate tribasic tetrahydrate, 98+%, for analysis

CAS: 5785-44-4 | C12H10Ca3O14·4H2O | 570.5 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 390082500
Price (USD)
35.10
Each
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
35.10
Each
Add to cart
Chemical Identifiers
CAS5785-44-4
IUPAC Nametricalcium bis(2-hydroxypropane-1,2,3-tricarboxylate) tetrahydrate
Molecular FormulaC12H18Ca3O18
InChI KeyLNIZKKFWMDARJV-UHFFFAOYSA-H
SMILESO.O.O.O.[Ca++].[Ca++].[Ca++].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Fluoride (F)=<0.003 %
Arsenic (As)=<0.0003 %
Lead (Pb)=<0.001 %
Appearance (Color)White
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link