4-Iodobutyronitrile, 97%
4-Iodobutyronitrile, 97%
4-Iodobutyronitrile, 97%
4-Iodobutyronitrile, 97%
Thermo Scientific Chemicals

4-Iodobutyronitrile, 97%

CAS: 6727-73-7 | C4H6IN | 195 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3903900101 gGlass bottle
Catalog number 390390010
Price (EUR)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6727-73-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.5350 to 1.5380 (20°C, 589 nm)
Appearance (Color)Clear light yellow
Infrared spectrumConforms
GC>=96.0 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0412283Certificate of AnalysisSep 22, 2023390390010
A0409168Certificate of AnalysisDec 13, 2022390390010
2 results displayed, search above for a specific certificate

Safety Data Sheets

Safety and Handling

Classification of the substance or mixture
CLP classification - Regulation(EC) No 1272/2008
Acute oral toxicity
Category 4
Acute dermal toxicity
Category 4
Acute Inhalation Toxicity - Dusts and Mists
Category 4
Label Elements
Signal Word

Warning

Hazard Statements

H302 + H312 + H332 - Harmful if swallowed, in contact with skin or if inhaled

EU Specific Hazard Statements

EUH032 - Contact with acids liberates very toxic gas

Precautionary Statements

P233 - Keep container tightly closed

P301 + P312 - IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell

P304 + P340 - IF INHALED: Remove person to fresh air and keep comfortable for breathing

P280 - Wear protective gloves/protective clothing/eye protection/face protection

P302 + P352 - IF ON SKIN: Wash with plenty of soap and water

Share catalog number, name or link

1x1 image pixel for data collection