tert-Butyl methyl ether, for analysis, Thermo Scientific Chemicals
tert-Butyl methyl ether, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

tert-Butyl methyl ether, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number 390480025
Price (USD)/ Each
-
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS1634-04-4
IUPAC Name2-methoxy-2-methylpropane
Molecular FormulaC5H12O
InChI KeyBZLVMXJERCGZMT-UHFFFAOYSA-N
SMILESCOC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.05 %
Residue after evaporation=<0.005 %
Barium (Ba)=<0.1 ppm
Chromium (Cr)=<0.02 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only