Thermo Scientific Chemicals

Di-n-butylamine, for HPLC, Thermo Scientific Chemicals

Catalog number: 390611000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

Di-n-butylamine, for HPLC, Thermo Scientific Chemicals

Catalog number: 390611000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 390611000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
111-92-2
IUPAC Name
dibutylamine
Molecular Formula
C8H19N
InChI Key
JQVDAXLFBXTEQA-UHFFFAOYSA-N
SMILES
CCCCNCCCC
Appearance (Color)
Clear colorless
GC
>=99.0 %
Water
=<0.1 %
Appearance (Form)
Liquid
UV transmittance
>=99 % (c=0.01M water, 254 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text