Thermo Scientific Chemicals

Calcium perchlorate tetrahydrate, pure, Thermo Scientific Chemicals

Catalog number: 390635000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Calcium perchlorate tetrahydrate, pure, Thermo Scientific Chemicals

Catalog number: 390635000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 390635000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
15627-86-8
Titration Complexometric
12.2 to 13.5 % (Calcium)
Appearance (Form)
Crystalline powder or crystals
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text