N,N-Diisopropylethylamine trihydrofluoride, 90%, Thermo Scientific Chemicals
N,N-Diisopropylethylamine trihydrofluoride, 90%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N,N-Diisopropylethylamine trihydrofluoride, 90%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Plastic bottle
Catalog number 390710010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS131600-43-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow-brown
Appearance (Form)Liquid
Infrared spectrumConforms
Titration with NaOH18 to 22 % (available HF)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only