Hexanes, for analysis, mixture of isomers, Thermo Scientific Chemicals
Hexanes, for analysis, mixture of isomers, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hexanes, for analysis, mixture of isomers, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 390740010
Price (USD)/ Each
35.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
42.20 
Save 6.55 (16%)
-
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
35.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
42.20 
Save 6.55 (16%)
Add to cart
Hexanes, for analysis, mixture of isomers, Thermo Scientific Chemicals
Catalog number390740010
Price (USD)/ Each
35.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
42.20 
Save 6.55 (16%)
-
Add to cart
Chemical Identifiers
IUPAC Namehexane
Molecular FormulaC6H14
InChI KeyVLKZOEOYAKHREP-UHFFFAOYSA-N
SMILESCCCCCC
Molecular Weight (g/mol)86.18
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<10 APHA
Substance darkened by H2SO4Passes test
Water=<0.01 %
Calcium (Ca)=<0.5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only