Cesium acetate, 99%, for analysis, Thermo Scientific Chemicals
Cesium acetate, 99%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cesium acetate, 99%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number 390831000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namecaesium(1+) acetate
Molecular FormulaC2H3CsO2
InChI KeyZOAIGCHJWKDIPJ-UHFFFAOYSA-M
SMILES[Cs+].CC([O-])=O
Molecular Weight (g/mol)191.95
View more
SpecificationsSpecification SheetSpecification Sheet
Potassium (K)=<20 ppm
Rubidium (Rb)=<0.12 %
Magnesium (Mg)=<3 ppm
Calcium (Ca)=<40 ppm
Aluminium (Al)=<6 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only