Sodium polyphosphate, pure
Sodium polyphosphate, pure
Sodium polyphosphate, pure
Sodium polyphosphate, pure
Sodium polyphosphate, pure
Thermo Scientific Chemicals

Sodium polyphosphate, pure

CAS: 50813-16-6 | (NaO3P)n
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
Packaging:
Plastic bottle
Catalog number 390930010
Price (USD)
66.65
Online Exclusive
74.10
Save 7.45 (10%)
Each
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
66.65
Online Exclusive
74.10
Save 7.45 (10%)
Each
Add to cart
Chemical Identifiers
CAS50813-16-6
IUPAC NameSodium polyphosphate
Molecular Formula(NaO3P)n
SMILES[Na]OP(-*)(=O)O-*
MDL NumberMFCD00134118
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Titration excess HCl65 to 70 %
Chloride (Cl)=<0.03 %
Sulfate (SO4)=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link