Thermo Scientific Chemicals

Lithium bromide, 4M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 390978000
800 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

Lithium bromide, 4M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 390978000
800 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 390978000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7550-35-8, 109-99-9
Appearance (Color)
Clear to light turbid colorless to
Appearance (Color)
yellow to pink
Appearance (Form)
Solution
Titration Argentometric
29 to 31 % (LiBr via Br)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text