3-Pyridinepropionic acid, 97%, Thermo Scientific Chemicals
3-Pyridinepropionic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Pyridinepropionic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
5 g
Packaging:
Glass bottle
Catalog number 390980100
Price (USD)/ Each
208.00
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
208.00
Add to cart
3-Pyridinepropionic acid, 97%, Thermo Scientific Chemicals
Catalog number390980100
Price (USD)/ Each
208.00
-
Add to cart
Chemical Identifiers
IUPAC Name3-(pyridin-3-yl)propanoic acid
Molecular FormulaC8H9NO2
InChI KeyWDGXIUUWINKTGP-UHFFFAOYSA-N
SMILESOC(=O)CCC1=CC=CN=C1
Molecular Weight (g/mol)151.17
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with NaOH>=96.0 %
Appearance (Form)Crystalline powder and/or chunks
Melting point153°C to 157°C
Appearance (Color)White to light beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only