1,1-Diphenylethylene oxide, Thermo Scientific Chemicals
1,1-Diphenylethylene oxide, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,1-Diphenylethylene oxide, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 391030050
Price (USD)/ Each
166.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
196.00 
Save 29.35 (15%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
166.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
196.00 
Save 29.35 (15%)
Add to cart
1,1-Diphenylethylene oxide, Thermo Scientific Chemicals
Catalog number391030050
Price (USD)/ Each
166.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
196.00 
Save 29.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2,2-diphenyloxirane
Molecular FormulaC14H12O
InChI KeyPRLJMHVNHLTQJJ-UHFFFAOYSA-N
SMILESC1OC1(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol)196.25
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point52°C to 57°C
Appearance (Form)Crystalline powder or crystals
NMR>=96.0 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only