N-Benzylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
N-Benzylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Benzylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 391040050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS29601-98-7
IUPAC NameN-benzylhydroxylamine hydrochloride
Molecular FormulaC7H10ClNO
InChI KeyYSNXOQGDHGUKCZ-UHFFFAOYSA-N
SMILESCl.ONCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Titration Argentometric97.5 to 102.5 %
Infrared spectrumConforms
Appearance (Color)White to off-white
Melting point108°C to 112°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only