N-Benzylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
N-Benzylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Benzylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number 391040250
Price (USD)/ Each
308.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
308.00
Add to cart
N-Benzylhydroxylamine hydrochloride, 98%, Thermo Scientific Chemicals
Catalog number391040250
Price (USD)/ Each
308.00
-
Add to cart
Chemical Identifiers
IUPAC NameN-benzylhydroxylamine hydrochloride
Molecular FormulaC7H10ClNO
InChI KeyYSNXOQGDHGUKCZ-UHFFFAOYSA-N
SMILESCl.ONCC1=CC=CC=C1
Molecular Weight (g/mol)159.61
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Melting point108°C to 112°C
Appearance (Form)Powder or crystals
Titration Argentometric97.5 to 102.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only