Thermo Scientific Chemicals

6-Bromo-2-picoline, 97%, Thermo Scientific Chemicals

Catalog number: 391050010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

6-Bromo-2-picoline, 97%, Thermo Scientific Chemicals

Catalog number: 391050010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 391050010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5315-25-3
IUPAC Name
2-bromo-6-methylpyridine
Molecular Formula
C6H6BrN
InChI Key
SOHDPICLICFSOP-UHFFFAOYSA-N
SMILES
CC1=CC=CC(Br)=N1
Refractive index
1.5620 to 1.5640 (20°C, 589 nm)
GC
>=96.0 %
Infrared spectrum
Conforms
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text