4-Chloromethylbenzonitrile, 98%, Thermo Scientific Chemicals
4-Chloromethylbenzonitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chloromethylbenzonitrile, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 391150100
Price (USD)/ Each
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name4-(chloromethyl)benzonitrile
Molecular FormulaC8H6ClN
InChI KeyLOQLDQJTSMKBJU-UHFFFAOYSA-N
SMILESClCC1=CC=C(C=C1)C#N
Molecular Weight (g/mol)151.59
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Crystalline powder
Appearance (Color)Off-white to light yellow
Melting point77°C to 80°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only