5-Cyanothiophene-2-boronic acid, 98%, Thermo Scientific Chemicals
5-Cyanothiophene-2-boronic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Cyanothiophene-2-boronic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 391270010
Price (USD)/ Each
172.00
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
172.00
Add to cart
5-Cyanothiophene-2-boronic acid, 98%, Thermo Scientific Chemicals
Catalog number391270010
Price (USD)/ Each
172.00
-
Add to cart
Chemical Identifiers
CAS305832-67-1
IUPAC Name(5-cyanothiophen-2-yl)boronic acid
Molecular FormulaC5H4BNO2S
InChI KeyZEOMEPSYIIQIND-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(S1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige to pink
Infrared spectrumConforms
Appearance (Form)Powder
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only