4-Iodopyridine, 96%, Thermo Scientific Chemicals
4-Iodopyridine, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Iodopyridine, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 391290050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS15854-87-2
IUPAC Name4-iodopyridine
Molecular FormulaC5H4IN
InChI KeyRTLUPHDWSUGAOS-UHFFFAOYSA-N
SMILESIC1=CC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals or needles or powder
GC>=95 %
Appearance (Color)Yellow to green to brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only