Citric acid, triammonium salt, +97%, pure, Thermo Scientific Chemicals
Citric acid, triammonium salt, +97%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Citric acid, triammonium salt, +97%, pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
50 g
250 g
Packaging:
Plastic bottle
Catalog number 391410010
Price (USD)/ Each
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS3458-72-8
IUPAC Nametriammonium 2-hydroxypropane-1,2,3-tricarboxylate
Molecular FormulaC6H17N3O7
InChI KeyYWYZEGXAUVWDED-UHFFFAOYSA-N
SMILES[NH4+].[NH4+].[NH4+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=97.0 %
Appearance (Color)White to off-white
Infrared spectrumConforms
Appearance (Form)Crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only