Thermo Scientific Chemicals

4-Amino-3-methoxybenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 391460010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Amino-3-methoxybenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 391460010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 391460010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2486-69-3
IUPAC Name
4-amino-3-methoxybenzoic acid
Molecular Formula
C8H9NO3
InChI Key
JNFGLYJROFAOQP-UHFFFAOYSA-N
SMILES
COC1=CC(=CC=C1N)C(O)=O
Appearance (Color)
White to light beige to grey
Infrared spectrum
Conforms
Appearance (Form)
Powder
Melting point
186°C to 191°C
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text