Chloro(triphenylphosphine)gold(I), 99%, Thermo Scientific Chemicals
Chloro(triphenylphosphine)gold(I), 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chloro(triphenylphosphine)gold(I), 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 mg
1 g
Packaging:
Glass bottle
Catalog number 391610050
Price (USD)/ Each
604.65
Online exclusive
672.00 
Save 67.35 (10%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
604.65
Online exclusive
672.00 
Save 67.35 (10%)
Add to cart
Chloro(triphenylphosphine)gold(I), 99%, Thermo Scientific Chemicals
Catalog number391610050
Price (USD)/ Each
604.65
Online exclusive
672.00 
Save 67.35 (10%)
-
Add to cart
Chemical Identifiers
CAS14243-64-2
IUPAC Namechlorogold; triphenylphosphane
Molecular FormulaC18H15AuClP
InChI KeyIFPWCRBNZXUWGC-UHFFFAOYSA-M
SMILESCl[Au].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Assay>=98.5 % (ex Au)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only