Thermo Scientific Chemicals

Indole-5-carboxaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: 391630050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Indole-5-carboxaldehyde, 98+%, Thermo Scientific Chemicals

Catalog number: 391630050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 391630050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1196-69-6
IUPAC Name
1H-indole-5-carbaldehyde
Molecular Formula
C9H7NO
InChI Key
ADZUEEUKBYCSEY-UHFFFAOYSA-N
SMILES
O=CC1=CC=C2NC=CC2=C1
Appearance (Form)
Powder
HPLC
>=98.0 %
Melting point
98°C to 103°C
Appearance (Color)
Orange to tan
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text