Thermo Scientific Chemicals

Ammonium thiosulfate, 96%, pure, Thermo Scientific Chemicals

Catalog number: 391665000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Ammonium thiosulfate, 96%, pure, Thermo Scientific Chemicals

Catalog number: 391665000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 391665000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7783-18-8
IUPAC Name
dihydroxy-1λ⁶-disulfen-1-one diamine
Molecular Formula
H8N2O3S2
InChI Key
XYXNTHIYBIDHGM-UHFFFAOYSA-N
SMILES
N.N.OS(O)(=O)=S
Appearance (Color)
White
Appearance (Form)
Crystalline powder or crystals and/or chunks
Titration Iodimetric
>=95 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text