4-Acetamidoacetophenone, 98%, Thermo Scientific Chemicals
4-Acetamidoacetophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Acetamidoacetophenone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 391820050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2719-21-3
IUPAC NameN-(4-acetylphenyl)acetamide
Molecular FormulaC10H11NO2
InChI KeyWECHHDJTILFYQT-UHFFFAOYSA-N
SMILESCC(=O)NC1=CC=C(C=C1)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point166°C to 170°C
Appearance (Form)Crystalline powder
Appearance (Color)Beige
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only