Thermo Scientific Chemicals

(3-Chloropropyl)triethoxysilane, 97+%, Thermo Scientific Chemicals

Catalog number: 391890010
1 kg, Glass bottle, Each
Thermo Scientific Chemicals

(3-Chloropropyl)triethoxysilane, 97+%, Thermo Scientific Chemicals

Catalog number: 391890010
1 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 391890010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5089-70-3
IUPAC Name
(3-chloropropyl)triethoxysilane
Molecular Formula
C9H21ClO3Si
InChI Key
KSCAZPYHLGGNPZ-UHFFFAOYSA-N
SMILES
CCO[Si](CCCCl)(OCC)OCC
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
GC
>=97.0 %
Appearance (Form)
Liquid
Refractive index
1.4190 to 1.4210 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text