Thermo Scientific Chemicals

(3-Isocyanatopropyl)triethoxysilane, 95%, Thermo Scientific Chemicals

Catalog number: 391901000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

(3-Isocyanatopropyl)triethoxysilane, 95%, Thermo Scientific Chemicals

Catalog number: 391901000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 391901000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
24801-88-5
IUPAC Name
triethoxy(3-isocyanatopropyl)silane
Molecular Formula
C10H21NO4Si
InChI Key
FRGPKMWIYVTFIQ-UHFFFAOYSA-N
SMILES
CCO[Si](CCCN=C=O)(OCC)OCC
GC
>=94 %
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless to yellow
Refractive index
1.4190 to 1.4240 (20°C, 589 nm)
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text