Magnesium phosphate, tribasic hydrate, 98+%, pure
Magnesium phosphate, tribasic hydrate, 98+%, pure
Magnesium phosphate, tribasic hydrate, 98+%, pure
Magnesium phosphate, tribasic hydrate, 98+%, pure
Thermo Scientific Chemicals

Magnesium phosphate, tribasic hydrate, 98+%, pure

CAS: 53408-95-0 | Mg3O8P2 | 262.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 391940010
Price (USD)
71.65
Special offer
Online exclusive
83.60
Save 11.95 (14%)
Each
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
71.65
Special offer
Online exclusive
83.60
Save 11.95 (14%)
Each
Add to cart
Chemical Identifiers
CAS53408-95-0
IUPAC Nametrimagnesium(2+) diphosphate
Molecular FormulaMg3O8P2
InChI KeyGVALZJMUIHGIMD-UHFFFAOYSA-H
SMILES[Mg++].[Mg++].[Mg++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Loss on ignition=<32.0 % (425°C)
Appearance (Color)White
Titration excess HCl98.0 to 102.0 % (On ignited basis)
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Kunden, die diesen Artikel ansahen, interessierten sich auch für



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0472738Certificate of AnalysisMay 19, 2025391942500, 391940010
A0471901Certificate of AnalysisMay 17, 2025391942500, 391940010
A0467444Certificate of AnalysisJan 31, 2025391942500, 391940010
A0463217Certificate of AnalysisMay 31, 2024391942500, 391940010
A0444152Certificate of AnalysisJan 11, 2024391942500, 391940010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection