1x1 image pixel for data collection
Magnesium phosphate, tribasic hydrate, 98+%, pure
Magnesium phosphate, tribasic hydrate, 98+%, pure
Magnesium phosphate, tribasic hydrate, 98+%, pure
Magnesium phosphate, tribasic hydrate, 98+%, pure
Thermo Scientific Chemicals

Magnesium phosphate, tribasic hydrate, 98+%, pure

CAS: 53408-95-0 | Mg3O8P2 | 262.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 391942500
Price (USD)
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS53408-95-0
IUPAC Nametrimagnesium(2+) diphosphate
Molecular FormulaMg3O8P2
InChI KeyGVALZJMUIHGIMD-UHFFFAOYSA-H
SMILES[Mg++].[Mg++].[Mg++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Loss on ignition=<32.0 % (425°C)
Appearance (Color)White
Titration excess HCl98.0 to 102.0 % (On ignited basis)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link