Thermo Scientific Chemicals

Dimethylethoxysilane, 94%, Thermo Scientific Chemicals

Catalog number: 392000050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Dimethylethoxysilane, 94%, Thermo Scientific Chemicals

Catalog number: 392000050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 392000050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14857-34-2
IUPAC Name
ethoxydimethylsilyl
Molecular Formula
C4H11OSi
InChI Key
DRUOQOFQRYFQGB-UHFFFAOYSA-N
SMILES
CCO[Si](C)C
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless
Refractive index
1.3670 to 1.3700 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=92 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text