Silicagel orange, for drying purposes, non toxic grade, Thermo Scientific Chemicals
Silicagel orange, for drying purposes, non toxic grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silicagel orange, for drying purposes, non toxic grade, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
Packaging:
View all
Glass Bottle
Catalog number 392030010
Price (USD)/ Each
96.65
Online exclusive
107.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
1 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
96.65
Online exclusive
107.00 
Save 10.35 (10%)
Add to cart
Silicagel orange, for drying purposes, non toxic grade, Thermo Scientific Chemicals
Catalog number392030010
Price (USD)/ Each
96.65
Online exclusive
107.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Beads
Water adsorption>=20 % (50% rH, 1 bar, 25°C)
ColorOrange (dry) to Colorless (wet)
Bead size2 to 5 mm
Appearance (Color)Orange
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Drying agent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only