Invitrogen™

IEF Marker 3-10

Catalog number: 3921201
Have Questions?
Invitrogen™

IEF Marker 3-10

Catalog number: 3921201
Have Questions?
Catalog Number
3921201
Unit Size
500 µL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
3921201500 µL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
IEF Marker 3-10 is a ready-to-use protein marker consisting of 13 purified isoforms for the use in IEF applications. The marker works well on all horizontal and vertical one-dimensional IEF gels and can be used in both native and denaturing conditions.

Compare and view all other protein standards and ladders ›

Applications
• Monitoring of separation on IEF gels

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Number of Markers
13
Product Type
IEF Marker
Ready to Load
Yes
Recommended Applications
IEF gels
Size Range
3.5 to 10.7 pI
Stain Type
Unstained
Gel Compatibility
Novex™ IEF Gels
Molecular Weight (g/mol)
10.7, 9.5, 8.3, 8.0, 7.8, 7.4, 6.9, 6.0, 5.3, 5.2, 4.5, 4.2, 3.5
Quantity
500 μL
Shipping Condition
Dry Ice
System Type
IEF

Contents & Storage

The IEF Marker 3-10 is supplied in 10% glycerol containing Bromophenol Blue (0.01%) and Methyl Red (0.01%).

Store in freezer (-5 to -30deg;C).

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text