Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)phenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 392151000
100 mL, AcroSeal™ Glass bottle, Each
Thermo Scientific Chemicals

3,5-Bis(trifluoromethyl)phenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 392151000
100 mL, AcroSeal™ Glass bottle, Each
Quantity
Packaging
Catalog number: 392151000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
112981-69-8
IUPAC Name
[3,5-bis(trifluoromethyl)phenyl](bromo)magnesium
Molecular Formula
C8H3BrF6Mg
InChI Key
FTKNONULORTDRS-UHFFFAOYSA-M
SMILES
FC(F)(F)C1=CC(=CC([Mg]Br)=C1)C(F)(F)F
Appearance (Color)
Dark brown
Appearance (Form)
Solution
Titration Grignard
0.40 to 0.60 M

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text