Thermo Scientific Chemicals

Trimethoxysilane, 95%, Thermo Scientific Chemicals

Catalog number: 392170050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Trimethoxysilane, 95%, Thermo Scientific Chemicals

Catalog number: 392170050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 392170050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2487-90-3
IUPAC Name
trimethoxysilyl
Molecular Formula
C3H9O3Si
InChI Key
PZJJKWKADRNWSW-UHFFFAOYSA-N
SMILES
CO[Si](OC)OC
Appearance (Color)
Clear colorless
Infrared spectrum
Conforms
Refractive index
1.3570 to 1.3600 (20°C, 589 nm)
GC
>=94 %
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text