Thermo Scientific Chemicals

Methyldiethoxysilane, 97%, Thermo Scientific Chemicals

Catalog number: 392260050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyldiethoxysilane, 97%, Thermo Scientific Chemicals

Catalog number: 392260050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 392260050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2031-62-1
IUPAC Name
diethoxy(methyl)silyl
Molecular Formula
C5H13O2Si
InChI Key
GAURFLBIDLSLQU-UHFFFAOYSA-N
SMILES
CCO[Si](C)OCC
Infrared spectrum
Conforms
Refractive index
1.3740 to 1.3760 (20°C, 589 nm)
GC
>=96.0 %
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text