4-Cyanophenylhydrazine hydrochloride, 97+%, Thermo Scientific Chemicals
4-Cyanophenylhydrazine hydrochloride, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Cyanophenylhydrazine hydrochloride, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 392300250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2863-98-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
HPLC>=97.0 %
Infrared spectrumConforms
Appearance (Color)Light orange to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only